iNOS Monoclonal Antibody (CXNFT), APC-eFluor™ 780, eBioscience™ - FAQs
iNOS Monoclonal Antibody (CXNFT), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for iNOS Monoclonal Antibody (CXNFT), APC-eFluor™ 780, eBioscience™ - FAQs (47-5920-80, 47-5920-82)