TER-119 Monoclonal Antibody (TER-119), APC-eFluor™ 780, eBioscience™ - FAQs
TER-119 Monoclonal Antibody (TER-119), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for TER-119 Monoclonal Antibody (TER-119), APC-eFluor™ 780, eBioscience™ - FAQs (47-5921-80, 47-5921-82)