Ly-6C Monoclonal Antibody (HK1.4), APC-eFluor™ 780, eBioscience™ - FAQs
Ly-6C Monoclonal Antibody (HK1.4), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Ly-6C Monoclonal Antibody (HK1.4), APC-eFluor™ 780, eBioscience™ - FAQs (47-5932-80, 47-5932-82)