TCR beta Monoclonal Antibody (H57-597), APC-eFluor™ 780, eBioscience™ - FAQs
TCR beta Monoclonal Antibody (H57-597), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for TCR beta Monoclonal Antibody (H57-597), APC-eFluor™ 780, eBioscience™ - FAQs (47-5961-82, 47-5961-80)