IgD Monoclonal Antibody (11-26c (11-26)), APC-eFluor™ 780, eBioscience™ - FAQs
IgD Monoclonal Antibody (11-26c (11-26)), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for IgD Monoclonal Antibody (11-26c (11-26)), APC-eFluor™ 780, eBioscience™ - FAQs (47-5993-80, 47-5993-82)