IL-13 Monoclonal Antibody (eBio13A), APC-eFluor™ 780, eBioscience™ - FAQs
IL-13 Monoclonal Antibody (eBio13A), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for IL-13 Monoclonal Antibody (eBio13A), APC-eFluor™ 780, eBioscience™ - FAQs (47-7133-82, 47-7133-80)