IL-17A Monoclonal Antibody (eBio64DEC17), APC-eFluor™ 780, eBioscience™ - FAQs
IL-17A Monoclonal Antibody (eBio64DEC17), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio64DEC17), APC-eFluor™ 780, eBioscience™ - FAQs (47-7179-42, 47-7179-41)