IFN gamma Monoclonal Antibody (XMG1.2), APC-eFluor™ 780, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (XMG1.2), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (XMG1.2), APC-eFluor™ 780, eBioscience™ - FAQs (47-7311-80, 47-7311-82)