IFN gamma Monoclonal Antibody (4S.B3), APC-eFluor™ 780, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (4S.B3), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (4S.B3), APC-eFluor™ 780, eBioscience™ - FAQs (47-7319-41, 47-7319-42)