Granzyme B Monoclonal Antibody (NGZB), APC-eFluor™ 780, eBioscience™ - FAQs
Granzyme B Monoclonal Antibody (NGZB), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for Granzyme B Monoclonal Antibody (NGZB), APC-eFluor™ 780, eBioscience™ - FAQs (47-8898-80, 47-8898-82)