KLRG1 Monoclonal Antibody (13F12F2), APC-eFluor™ 780, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (13F12F2), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (13F12F2), APC-eFluor™ 780, eBioscience™ - FAQs (47-9488-42, 47-9488-41)