HELIOS Monoclonal Antibody (22F6), APC-eFluor™ 780, eBioscience™ - FAQs
HELIOS Monoclonal Antibody (22F6), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for HELIOS Monoclonal Antibody (22F6), APC-eFluor™ 780, eBioscience™ - FAQs (47-9883-42, 47-9883-41)