HLA-DR Monoclonal Antibody (LN3), APC-eFluor™ 780, eBioscience™ - FAQs
HLA-DR Monoclonal Antibody (LN3), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for HLA-DR Monoclonal Antibody (LN3), APC-eFluor™ 780, eBioscience™ - FAQs (47-9956-41, 47-9956-42)