CD279 (PD-1) Monoclonal Antibody (J43), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD279 (PD-1) Monoclonal Antibody (J43), APC-eFluor™ 780, eBioscience™ - FAQs (47-9985-80, 47-9985-82)