Finnpipette™ R&D Systems GLP Kits
Thermo Scientific™

Finnpipette™ R&D Systems GLP Kits

A Novus product, part of R&D Systems. Access everything needed for pipetting in these all-inclusive GLP kits featuring ergonomic and easy-to-use pipetters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
470047010 to 1000 μL
4700490100 to 5000 μL
47011201 to 1000 μL
470113010 to 10,000 μL
Catalog number 4700470
Price (USD)
983.52
Online exclusive
1,677.80
Save 694.28
Each
Volume (Metric):
10 to 1000 μL
Price (USD)
983.52
Online exclusive
1,677.80
Save 694.28
Each

Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ R&D Systems GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ R&D Systems™ pipetters, appropriate stands and sample tips.

R&D Systems GLP Kit 1 includes:

  • Finnpipette R&D Systems 10-100μL
  • Finnpipette R&D Systems 100-1000μL
  • Two Finnpipette R&D Systems stands
  • One rack of Finntip Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Finnpipette R&D Systems brochure

R&D Systems GLP Kit 2 includes:

  • Finnpipette R&D Systems 100-1000μL
  • Finnpipette R&D Systems 0.5-5mL
  • Two Finnpipette R&D Systems stands
  • One rack of Finntip 5ml and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Finnpipette R&D Systems brochure
  • Pipetting Guide

R&D Systems GLP Kit 3 includes:

  • R&D Systems pipette, 1-10μL, micro
  • R&D Systems pipette, 10-100μL
  • R&D Systems pipette, 100-1000μL
  • Three Finnpipette R&D Systems pipette stands
  • One rack of Flex 10, Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • R&D Systems brochure

R&D Systems GLP Kit 4 includes:

  • R&D Systems pipette, 10-100μL
  • R&D Systems pipette, 100-1000μL
  • R&D Systems pipette, 1-10mL
  • Three Finnpipette R&D Systems pipette stands
  • One rack of Flex 200, Flex 1000 and Finntip 10ml
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • R&D Systems brochure
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
IncludesNovus 100 and 1000 μL single channel pipettes, two Novus stands, two sample Finntip racks
ModelNovus
ColorAssorted
Number of Channels1
Product LineNovus
TypeGLP Kit
Volume (Metric)10 to 1000 μL
Unit SizeEach