Finnpipette™ Novus GLP Kits
Thermo Scientific™

Finnpipette™ Novus GLP Kits

Access everything needed for pipetting in these all-inclusive GLP kits featuring ergonomic and easy-to-use pipetters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
4700490100 to 5000 μL
470047010 to 1000 μL
47011201 to 1000 μL
470113010 to 10,000 μL
Catalog number 4700490
Price (USD)
1,404.96
Online exclusive
1,677.80
Save 272.84
Each
Volume (Metric):
100 to 5000 μL
Price (USD)
1,404.96
Online exclusive
1,677.80
Save 272.84
Each

Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ Novus GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.

Novus GLP Kit 1 includes:

  • Finnpipette Novus 10-100μL
  • Finnpipette Novus 100-1000μL
  • Two Finnpipette Novus stands
  • One rack of Finntip Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Finnpipette Novus brochure

Novus GLP Kit 2 includes:

  • Finnpipette Novus 100-1000μL
  • Finnpipette Novus 0.5-5mL
  • Two Finnpipette Novus stands
  • One rack of Finntip 5ml and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Finnpipette Novus brochure
  • Pipetting Guide

Novus GLP Kit 3 includes:

  • Novus pipette, 1-10μL, micro
  • Novus pipette, 10-100μL
  • Novus pipette, 100-1000μL
  • Three Finnpipette Novus pipette stands
  • One rack of Flex 10, Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Novus brochure

Novus GLP Kit 4 includes:

  • Novus pipette, 10-100μL
  • Novus pipette, 100-1000μL
  • Novus pipette, 1-10mL
  • Three Finnpipette Novus pipette stands
  • One rack of Flex 200, Flex 1000 and Finntip 10ml
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Novus brochure
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
IncludesNovus 1000 and 5000 μL single channel pipettes, two Novus stands, two sample Finntip racks
ModelNovus
ColorAssorted
Number of Channels1
Product LineNovus
TypeGLP Kit
Volume (Metric)100 to 5000 μL
Unit SizeEach