Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
Thermo Scientific™

Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits

Enjoy the convenience of having a variety of Finnpipette™ F1 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호부피(미터법)
4700850N1 to 1000 μL
4700860N0.2 to 1000 μL
4700865N10 to 10,000 μL
4701060N2 to 1000 μL
카탈로그 번호 4700850N
제품 가격(KRW)
933,000
온라인 행사
Ends: 31-Dec-2026
1,097,000
할인액 164,000 (15%)
Each
부피(미터법):
1 to 1000 μL
제품 가격(KRW)
933,000
온라인 행사
Ends: 31-Dec-2026
1,097,000
할인액 164,000 (15%)
Each
Support good laboratory pipetting in your lab with the Thermo Scientific™ Finnpipette™ F1 GLP Kits, which offer a variety of single channels in a range of volumes that are packaged with compatible Finntips, and a convenient pipette stand. Select the kit that is right for you, and take advantage of the Finnpipette F1 performance and lightweight design to maximize your comfort.

For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.

Finnpipette F1: Everything you want in your everyday, go-to pipette

Features:

  • Lightweight handle design
  • Low plunger and tip attachment & ejection forces
  • Secure volume lock to prevent undesirable volume drift during operation
  • Silent, effortless volume adjustment
  • Adjustable finger rest - comfort for both right- and left-handed users
  • Color coded together with Finntips for easier recognition of compatible tips

Kit 1 includes:

  • Three Thermo Scientific Finnpipette F1 Pipettes from 1 to 1000 μL:
    • 1-10 μL, micro
    • 10-100 μL
    • 100-1000 μL
  • Grip Strip for each handle to improve grip
  • Flex tips: 10 micro: 1 x 96; 200: 1 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette F1 Pipettes from 0.2 to 1000 μL:
    • 0.2-2 μL
    • 2-20 μL univ
    • 20-200 μL
    • 100-1000 μL
  • Grip Strip for each handle to improve grip
  • Flex tips: 10 micro: 1 x 96; 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette F1 Pipettes from 10 to 10,000 μL:
    • 10-100 μL
    • 100-1000 μL
    • 1000-10,000 μL
  • Grip Strip for each handle to improve grip
  • Flex tips: 200: 1 x 96; 1000: 1 x 96; FT 10 mL: 1 x 24
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette F1 Pipettes from 20 to 1000 μL:
    • 2-20 μL
    • 20-200 μL
    • 100-1000 μL 
  • Grip Strip for each handle to improve grip
  • Flex tips: 200μL: 2 x 96; 1000 μL: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

The kit contains samples racks of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains helpful tips for pipetting including pipetting techniques, ergonomic considerations and how to select tips for best performance.

Warranty: 5 years with web registration.

"

사양
함께 사용가능한 팁Finntip Flex tips: 10; 200; 1000
설명Finnpipette F1 GLP Kit 1: 1-10μL micro, 10-100μL and 100-1000μL
채널 수1
유형GLP Kit
부피(미터법)1 to 1000 μL
Unit SizeEach