Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Thermo Scientific™

Immuno Standard Modules White and Black

Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WellSurface Treatment
475515Black400 μLMaxiSorp
437591White350 μLMaxiSorp
437702White350 μLPolySorp
437915Clear350 μLMaxiSorp
475523Black400 μLPolySorp
7566White380 μLHydrophobic
Catalog number 475515
Price (USD)
694.26
Online exclusive
809.97
Save 115.71
Case of 60
Color:
Black
Volume (Metric) Well:
400 μL
Surface Treatment:
MaxiSorp
Price (USD)
694.26
Online exclusive
809.97
Save 115.71
Case of 60
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.

Features:

  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Binding PropertyHydrophilic
Binding TypeAdsorption
BreakapartNo
CertifiedYes
Detection MethodFluorescence
For Use With (Application)Fluorescence
FormatStandard
No. per Case60
No. per Pack5
Packaging60 Cases
Plate BlockingNone
Product TypeMicroplate Strip
Target MoleculeAntibodies (Glycoproteins)
Volume (Metric) Well400 μL
Volume (Metric) Working350 μL
Array8 x 12
ColorBlack
MaterialPolystyrene
No. of Wells96
Surface TreatmentMaxiSorp
Well ShapeRound, Flat Bottom (F-well)
Unit SizeCase of 60

Frequently asked questions (FAQs)

What length of peptide is ideal for binding to the Thermo Scientific MaxiSorp surface? What are the detection limitations?

We have tested and found that a 3 amino acid peptide (Pro, Leu, Gly) cannot be detected when passively adsorbed on the MaxiSorp surface. However, this peptide can be detected when covalently immobilized using CovaLink NH Modules and CovaLink NH2 Modules and Plates. Using covalent immobilization of small peptide residues, you can typically obtain a better orientation of the molecule and reduced problems with antibody recognition of the peptide due to masking of the epitope. We have discovered that a 7 amino acid peptide from the MHC Class II antigen can be detected when adsorbed on the MaxiSorp surface. We state that the detection limitation using the MaxiSorp surface is between 3 and 7 amino acid residues.
One additional note is that detection is contingent upon the orientation of the peptide when immobilized. If the active site is inactivated or hidden at the site facing the solid phase, no detection signal is observed.

Can I bind either single- or double-stranded DNA to the Thermo Scientific MaxiSorp surface?

Single-stranded DNA can be adsorbed to our MaxiSorp surface using approximately 10 µg ssDNA per mL PBS, pH 8.2, although the stability is uncertain. Based on our experience, ssDNA immobilized on the MaxiSorp surface is so loosely bound that it can be removed by stringent washing.
Double-stranded DNA will not bind to the MaxiSorp surface. DNA, however, can be covalently bound to Nunc NucleoLink Strips.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

What is the difference between certified and non-certified MaxiSorp plates and modules?

Both of these surfaces are identical. The only difference between them is that for the certified plates, a representative sample from each manufacturing lot undergoes a Binding Capacity test. This test is an ELISA-like assay used in our quality control laboratories to ensure binding capabilities.