CD1d Monoclonal Antibody (1B1), eFluor™ 450, eBioscience™ - FAQs
CD1d Monoclonal Antibody (1B1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD1d Monoclonal Antibody (1B1), eFluor™ 450, eBioscience™ - FAQs (48-0011-80, 48-0011-82)