CD1a Monoclonal Antibody (HI149), eFluor™ 450, eBioscience™ - FAQs
CD1a Monoclonal Antibody (HI149), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD1a Monoclonal Antibody (HI149), eFluor™ 450, eBioscience™ - FAQs (48-0019-41, 48-0019-42)