CD2 Monoclonal Antibody (RPA-2.10), eFluor™ 450, eBioscience™ - FAQs
CD2 Monoclonal Antibody (RPA-2.10), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD2 Monoclonal Antibody (RPA-2.10), eFluor™ 450, eBioscience™ - FAQs (48-0029-42, 48-0029-41)