CD3e Monoclonal Antibody (145-2C11), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD3e Monoclonal Antibody (145-2C11), eFluor™ 450, eBioscience™ - FAQs (48-0031-82, 48-0031-80)