CD3e Monoclonal Antibody (eBio500A2 (500A2)), eFluor™ 450, eBioscience™ - FAQs
CD3e Monoclonal Antibody (eBio500A2 (500A2)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD3e Monoclonal Antibody (eBio500A2 (500A2)), eFluor™ 450, eBioscience™ - FAQs (48-0033-82, 48-0033-80)