CD3 Monoclonal Antibody (SK7), eFluor™ 450, eBioscience™ - FAQs
CD3 Monoclonal Antibody (SK7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (SK7), eFluor™ 450, eBioscience™ - FAQs (48-0036-42, 48-0036-41)