CD3 Monoclonal Antibody (UCHT1), eFluor™ 450, eBioscience™ - FAQs
CD3 Monoclonal Antibody (UCHT1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (UCHT1), eFluor™ 450, eBioscience™ - FAQs (48-0038-80, 48-0038-82)