CD4 Monoclonal Antibody (OKT4 (OKT-4)), eFluor™ 450, eBioscience™ - FAQs
CD4 Monoclonal Antibody (OKT4 (OKT-4)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (OKT4 (OKT-4)), eFluor™ 450, eBioscience™ - FAQs (48-0048-42, 48-0048-41)