CD4 Monoclonal Antibody (RPA-T4), eFluor™ 450, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RPA-T4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RPA-T4), eFluor™ 450, eBioscience™ - FAQs (48-0049-42, 48-0049-41)