CD5 Monoclonal Antibody (53-7.3), eFluor™ 450, eBioscience™ - FAQs
CD5 Monoclonal Antibody (53-7.3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD5 Monoclonal Antibody (53-7.3), eFluor™ 450, eBioscience™ - FAQs (48-0051-80, 48-0051-82)