CD8a Monoclonal Antibody (53-6.7), eFluor™ 450, eBioscience™ - FAQs
CD8a Monoclonal Antibody (53-6.7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (53-6.7), eFluor™ 450, eBioscience™ - FAQs (48-0081-82, 48-0081-80)