CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), eFluor™ 450, eBioscience™ - FAQs
CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), eFluor™ 450, eBioscience™ - FAQs (48-0083-80, 48-0083-82)