CD8a Monoclonal Antibody (RPA-T8), eFluor™ 450, eBioscience™ - FAQs
CD8a Monoclonal Antibody (RPA-T8), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (RPA-T8), eFluor™ 450, eBioscience™ - FAQs (48-0088-42, 48-0088-41)