CD11c Monoclonal Antibody (N418), eFluor™ 450, eBioscience™ - FAQs
CD11c Monoclonal Antibody (N418), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (N418), eFluor™ 450, eBioscience™ - FAQs (48-0114-82, 48-0114-80)