CD15 Monoclonal Antibody (HI98), eFluor™ 450, eBioscience™ - FAQs
CD15 Monoclonal Antibody (HI98), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD15 Monoclonal Antibody (HI98), eFluor™ 450, eBioscience™ - FAQs (48-0159-41, 48-0159-42)