CD16/CD32 Monoclonal Antibody (93), eFluor™ 450, eBioscience™ - FAQs
CD16/CD32 Monoclonal Antibody (93), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD16/CD32 Monoclonal Antibody (93), eFluor™ 450, eBioscience™ - FAQs (48-0161-80, 48-0161-82)