CD16 Monoclonal Antibody (eBioCB16 (CB16)), eFluor™ 450, eBioscience™ - FAQs
CD16 Monoclonal Antibody (eBioCB16 (CB16)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD16 Monoclonal Antibody (eBioCB16 (CB16)), eFluor™ 450, eBioscience™ - FAQs (48-0168-42, 48-0168-41)