CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 450, eBioscience™ - FAQs
CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 450, eBioscience™ - FAQs (48-0193-82, 48-0193-80)