CD19 Monoclonal Antibody (SJ25C1), eFluor™ 450, eBioscience™ - FAQs
CD19 Monoclonal Antibody (SJ25C1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (SJ25C1), eFluor™ 450, eBioscience™ - FAQs (48-0198-42, 48-0198-41)