CD19 Monoclonal Antibody (HIB19), eFluor™ 450, eBioscience™ - FAQs
CD19 Monoclonal Antibody (HIB19), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (HIB19), eFluor™ 450, eBioscience™ - FAQs (48-0199-41, 48-0199-42)