CD20 Monoclonal Antibody (2H7), eFluor™ 450, eBioscience™ - FAQs
CD20 Monoclonal Antibody (2H7), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD20 Monoclonal Antibody (2H7), eFluor™ 450, eBioscience™ - FAQs (48-0209-41, 48-0209-42)