CD23 Monoclonal Antibody (EBVCS2), eFluor™ 450, eBioscience™ - FAQs
CD23 Monoclonal Antibody (EBVCS2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD23 Monoclonal Antibody (EBVCS2), eFluor™ 450, eBioscience™ - FAQs (48-0238-42)