CD24 Monoclonal Antibody (M1/69), eFluor™ 450, eBioscience™ - FAQs
CD24 Monoclonal Antibody (M1/69), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD24 Monoclonal Antibody (M1/69), eFluor™ 450, eBioscience™ - FAQs (48-0242-80, 48-0242-82)