CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), eFluor™ 450, eBioscience™ - FAQs
CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), eFluor™ 450, eBioscience™ - FAQs (48-0247-41, 48-0247-42)