CD25 Monoclonal Antibody (eBio3C7 (3C7)), eFluor™ 450, eBioscience™ - FAQs
CD25 Monoclonal Antibody (eBio3C7 (3C7)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (eBio3C7 (3C7)), eFluor™ 450, eBioscience™ - FAQs (48-0253-80, 48-0253-82)