CD25 Monoclonal Antibody (CD25-4E3), eFluor™ 450, eBioscience™ - FAQs
CD25 Monoclonal Antibody (CD25-4E3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (CD25-4E3), eFluor™ 450, eBioscience™ - FAQs (48-0257-42)