CD25 Monoclonal Antibody (BC96), eFluor™ 450, eBioscience™ - FAQs
CD25 Monoclonal Antibody (BC96), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (BC96), eFluor™ 450, eBioscience™ - FAQs (48-0259-41, 48-0259-42)