CD27 Monoclonal Antibody (O323), eFluor™ 450, eBioscience™ - FAQs
CD27 Monoclonal Antibody (O323), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD27 Monoclonal Antibody (O323), eFluor™ 450, eBioscience™ - FAQs (48-0279-42, 48-0279-41)