CD31 (PECAM-1) Monoclonal Antibody (390), eFluor™ 450, eBioscience™ - FAQs
CD31 (PECAM-1) Monoclonal Antibody (390), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD31 (PECAM-1) Monoclonal Antibody (390), eFluor™ 450, eBioscience™ - FAQs (48-0311-82, 48-0311-80)