CD32 Monoclonal Antibody (6C4 (CD32)), eFluor™ 450, eBioscience™ - FAQs
CD32 Monoclonal Antibody (6C4 (CD32)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD32 Monoclonal Antibody (6C4 (CD32)), eFluor™ 450, eBioscience™ - FAQs (48-0329-41, 48-0329-42)