SIGLEC H Monoclonal Antibody (eBio440c), eFluor™ 450, eBioscience™ - FAQs
SIGLEC H Monoclonal Antibody (eBio440c), eFluor™ 450, eBioscience™ - FAQs
View additional product information for SIGLEC H Monoclonal Antibody (eBio440c), eFluor™ 450, eBioscience™ - FAQs (48-0333-80, 48-0333-82)