CD33 Monoclonal Antibody (P67.6), eFluor™ 450, eBioscience™ - FAQs
CD33 Monoclonal Antibody (P67.6), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD33 Monoclonal Antibody (P67.6), eFluor™ 450, eBioscience™ - FAQs (48-0337-41, 48-0337-42)